missing translation for 'onlineSavingsMsg'
Learn More

Sodium tin(IV) oxide trihydrate, 96%

Product Code. 11498527
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
Unit Size:
100g
2500g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11498527 500 g 500g
11488527 100 g 100g
11408537 2500 g 2500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11498527 Supplier Thermo Scientific Alfa Aesar Supplier No. A18611.36
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 12209-98-2 | H6Na2O6Sn | 266.732 g/mol

Sodium tin(IV) oxide trihydrate is used in the preparation of boscage-like stannic oxide by facile one-step hydrothermal route.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium tin(IV) oxide trihydrate is used in the preparation of boscage-like stannic oxide by facile one-step hydrothermal route.

Solubility
Soluble in water. Insoluble in ethanol and acetone.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12209-98-2
Molecular Formula H6Na2O6Sn
Molecular Weight (g/mol) 266.732
MDL Number MFCD00150189
InChI Key SFXJSNATBHJIDS-UHFFFAOYSA-N
Synonym sodium stannate trihydrate, unii-nj7c1v83kg, nj7c1v83kg, sodium stannate inci, sodium stannate vandf, disodium stannate, trihydrate, sodium stannate iv trihydrate mi, 2na.so3n.3h2o, sodiumstannatetrihydrate, sodium tin iv oxide trihydrate
PubChem CID 21977542
IUPAC Name disodium;dioxido(oxo)tin;trihydrate
SMILES O.O.O.[O-][Sn](=O)[O-].[Na+].[Na+]

Specifications

Quantity 500 g
Merck Index 14,8677
Solubility Information Soluble in water. Insoluble in ethanol and acetone.
Formula Weight 266.75 (212.67 Anhydrous)
Percent Purity 98%
Odor Odorless
Chemical Name or Material Sodium tin(IV) oxide trihydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.