missing translation for 'onlineSavingsMsg'
Learn More

Sodium tin(IV) oxide trihydrate, 96%

Product Code. 11408537
Click to view available options
Quantity:
100 g
500 g
2500 g
Unit Size:
100g
2500g
500g
This item is not returnable. View return policy

Product Code. 11408537

Brand: Thermo Scientific Alfa Aesar A18611.0E

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 12209-98-2 | H6Na2O6Sn | 266.732 g/mol

Sodium tin(IV) oxide trihydrate is used in the preparation of boscage-like stannic oxide by facile one-step hydrothermal route.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium tin(IV) oxide trihydrate is used in the preparation of boscage-like stannic oxide by facile one-step hydrothermal route.

Solubility
Soluble in water. Insoluble in ethanol and acetone.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12209-98-2
Molecular Formula H6Na2O6Sn
Molecular Weight (g/mol) 266.732
MDL Number MFCD00150189
InChI Key SFXJSNATBHJIDS-UHFFFAOYSA-N
Synonym sodium stannate trihydrate, unii-nj7c1v83kg, nj7c1v83kg, sodium stannate inci, sodium stannate vandf, disodium stannate, trihydrate, sodium stannate iv trihydrate mi, 2na.so3n.3h2o, sodiumstannatetrihydrate, sodium tin iv oxide trihydrate
PubChem CID 21977542
IUPAC Name disodium;dioxido(oxo)tin;trihydrate
SMILES O.O.O.[O-][Sn](=O)[O-].[Na+].[Na+]

Specifications

Quantity 2500 g
Merck Index 14,8677
Solubility Information Soluble in water. Insoluble in ethanol and acetone.
Formula Weight 266.75 (212.67 Anhydrous)
Percent Purity 98%
Odor Odorless
Chemical Name or Material Sodium tin(IV) oxide trihydrate

RUO – Research Use Only

Corrección del contenido de un producto

Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.

Título del producto

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.

Gracias por ayudarnos a mejorar nuestra web. Su comentario ha sido enviado