missing translation for 'onlineSavingsMsg'
Learn More

Sodium thiosulphate 0.1 mol/L (0.1 N) in aqueous solution, BAKER ANALYZED™, J.T. Baker™

Product Code. 10698331 Shop All J.T.Baker Products
Change view
Click to view available options
Quantity:
1 L
10 L
Unit Size:
1 L
10 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10698331 10 L 10 L
10526622 1 L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10698331 Supplier JT Baker Supplier No. 7101.9010
  • ONLINE EXCLUSIVE PRICE
    £298.20
    £168.23 / 10 L
    Save £129.97
    43% Off
 Restricted Item
Estimated Shipment: 12-02-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Laboratory Reagent

Chemical Identifiers

CAS 7772-98-7
Molecular Formula Na2O3S2
Molecular Weight (g/mol) 158.10
MDL Number MFCD00003499
InChI Key AKHNMLFCWUSKQB-UHFFFAOYSA-L
Synonym Sodium hyposulphite , di-Sodium thiosulphate
IUPAC Name disodium pentahydrate sulfanidesulfonate
SMILES [Na+].[Na+].[O-]S([S-])(=O)=O

Especificaciones

Quantity 10 L
Concentration or Composition (by Analyte or Components) 0.0997 - 0.1003 mol/L
Packaging Plastic container
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.