missing translation for 'onlineSavingsMsg'
Learn More

Sodium thiosulphate 0.1 mol/L (0.1 N) in aqueous solution, BAKER ANALYZED™, J.T. Baker™

Product Code. 10526622 Shop All J.T.Baker Products
Change view
Click to view available options
Quantity:
1 L
10 L
Unit Size:
1 L
10 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10526622 1 L 1 L
10698331 10 L 10 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10526622 Supplier JT Baker Supplier No. 7101.1000
  • ONLINE EXCLUSIVE PRICE
    £203.00
    £115.10 / 1 L
    Save £87.90
    43% Off
 Restricted Item
Estimated Shipment: 28-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Laboratory Reagent

Chemical Identifiers

CAS 7772-98-7
Molecular Formula Na2O3S2
Molecular Weight (g/mol) 158.10
MDL Number MFCD00003499
InChI Key AKHNMLFCWUSKQB-UHFFFAOYSA-L
Synonym Sodium hyposulphite , di-Sodium thiosulphate
IUPAC Name disodium pentahydrate sulfanidesulfonate
SMILES [Na+].[Na+].[O-]S([S-])(=O)=O

Specifications

Quantity 1 L
Concentration or Composition (by Analyte or Components) 0.0997 - 0.1003 mol/L
Packaging Plastic bottle
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.