missing translation for 'onlineSavingsMsg'
Learn More

Sodium thiosulfate solution, Volumetric, Reag. Ph. Eur., 0.1 M Na2S2O3 (0.1N), Honeywell Fluka™

Product Code. 15654870 Shop All Honeywell Fluka Products
Click to view available options
Quantity:
1 L
5 L
10 L
6 x 1 L
Unit Size:
1 L
10 L
5 L
Pack of 6
This item is not returnable. View return policy

Product Code. 15654870

Brand: Honeywell-Fluka 3524510L

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Volumetric, Reag. Ph. Eur., 0.1 M Na2S2O3 (0.1N)

Honeywell´s high-quality titration products cover the complete titration workflow and include:

  • Solutions for volumetric titration (acids, bases, or salts)
  • Buffers available as concentrates or ready-to-use

 

  • Reagents for complexometry:
    • Aminopolycarboxylic acids (EDTA/NTA analogs)
    • Masking agents
  • Indicators for volumetric and complexometric titrations

 


 

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7772-98-7
Molecular Formula Na2O3S2
Molecular Weight (g/mol) 158.10
MDL Number MFCD00003499
InChI Key AKHNMLFCWUSKQB-UHFFFAOYSA-L
Synonym sodium thiosulfate, sodium thiosulphate, disodium thiosulfate, sodium thiosulfate anhydrous, hypo, sodiumthiosulfate, chlorine cure, chlorine control, declor-it, thiosulfuric acid, disodium salt
PubChem CID 24477
SMILES [Na+].[Na+].[O-]S([S-])(=O)=O

Specifications

Quantity 10 L
Linear Formula Na2S2O3
UN Number NONH for all modes of transport
IUPAC Name disodium sulfanidesulfonate
Formula Weight 158.11
Grade Reagent Ph. Eur.
Chemical Name or Material Sodium thiosulfate solution
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.