missing translation for 'onlineSavingsMsg'
Learn More

Sodium Thiosulfate Solution 0.1M (0.1N), NIST Standard Concentrate, for Volumetric Analysis, Fisher Chemical

Product Code. 10677412 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
6 AMP
Packaging:
Ampoule
Unit Size:
6 ampoules
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10677412 6 AMP Ampoule 6 ampoules
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10677412 Supplier Fisher Chemical Supplier No. J/7950C/90
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

6AMP, Ampoule

TRUSTED_SUSTAINABILITY

Specifications

Boiling Point 100°C
Color Colorless
Physical Form Liquid
Quantity 6 AMP
MDL Number 3499
Packaging Ampoule
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.