missing translation for 'onlineSavingsMsg'
Learn More

Sodium thiosulfate pentahydrate, ACS, 99.5-101.0%, Thermo Scientific Chemicals

Catalog Number 11372468
Click to view available options
Quantity:
500 g
2 kg
10102-17-7
H10Na2O8S2
248.172
MFCD00149186
PODWXQQNRWNDGD-UHFFFAOYSA-L
sodium thiosulfate pentahydrate, ametox, sodium thiosulfate, pentahydrate, antichlor, tinver, disodium thiosulfate pentahydrate, unii-hx1032v43m, ccris 3952, thiosulfuric acid, disodium salt, pentahydrate, sodium hyposulfite pentahydrate
61475
CHEBI:32150
disodium;dioxido-oxo-sulfanylidene-$l^{6}-sulfane;pentahydrate
O.O.O.O.O.[O-]S(=O)(=S)[O-].[Na+].[Na+]
10102-17-7
H10Na2O8S2
248.172
MFCD00149186
PODWXQQNRWNDGD-UHFFFAOYSA-L
sodium thiosulfate pentahydrate, ametox, sodium thiosulfate, pentahydrate, antichlor, tinver, disodium thiosulfate pentahydrate, unii-hx1032v43m, ccris 3952, thiosulfuric acid, disodium salt, pentahydrate, sodium hyposulfite pentahydrate
61475
CHEBI:32150
disodium;dioxido-oxo-sulfanylidene-$l^{6}-sulfane;pentahydrate
O.O.O.O.O.[O-]S(=O)(=S)[O-].[Na+].[Na+]

Maximum level of impurities: Insoluble matter 0.005%, pH of a 5% solution 6.0-8.4 at 25°, Nitrogen compounds (as N) 0.002%, Sulfate and sulfite (as SO4) 0.1%, Sulfide P.T. (limit about 1ppm)

Sodium thiosulfate pentahydrate is used as an analytical reagent in laboratories. It is used in photographic processing and in gold extraction. Its alkylated derivatives, S-alkylthiosulfonates, are known as Bunte salts. It is involved in the preparation of thioglycolic acid. Furthermore, it is used to dechlorinate tap water, for removing free bromine from solutions and to check the pH of bleach solutions with liquid indicators. Other uses include tanning of leather; as an active component in hand warmers and chemical heating pads and as an anionic surfactant to aid in the dispersion of pharmaceutical preparations.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Boiling Point 100°C-5H{2}O
Melting Point 48°C
Color Colorless
Physical Form Crystalline
Quantity 500 g
Assay Percent Range 99.5-101.0%
Solubility Information Soluble in water and ammonia. Insoluble in alcohol and organic solvents.
Formula Weight 248.17 (158.10 Anhydrous)
Grade ACS
Odor Odorless
Appearance Colorless crystalline
Density 1.69 g/cm3
Chemical Name or Material Sodium thiosulfate pentahydrate, For ACS analysis
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'