missing translation for 'onlineSavingsMsg'
Learn More

Sodium thiosulfate concentrate, For 1L standard solution, 0.1 M Na2S2O3 (0.1N), Solstice

Product Code. 15655240
Change view
Click to view available options
Quantity:
1 Pc.
6 x 1 Ea.
Unit Size:
Each
Pack of 6
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15655240 6 x 1 Ea. Pack of 6
15645240 1 Pc. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15655240 Supplier Fluka Supplier No. 382006X1EA
 Restricted Item
Estimated Shipment: 21-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

for 1L standard solution, 0.1 M Na2S2O3 (0.1N)

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10102-17-7 , 6132-02-1 , 7732-18-5
MDL Number MFCD00003499
ChEBI CHEBI:32150

Specifications

Boiling Point 100°C
CAS Min % ≥20.0000%
CAS Max % ≤40.0000%, ≤0.1000%, ≤59.9000%
Physical Form Concentrate
Quantity 6 x 1 Ea.
Packaging Ampoule
Molecular Formula H10Na2O8S2
Linear Formula Na2S2O3
UN Number NONH for all modes of transport
Synonym sodium thiosulfate pentahydrate, ametox, sodium thiosulfate, pentahydrate, antichlor, tinver, disodium thiosulfate pentahydrate, unii-hx1032v43m, ccris 3952, thiosulfuric acid, disodium salt, pentahydrate, sodium hyposulfite pentahydrate
InChI Key PODWXQQNRWNDGD-UHFFFAOYSA-L
SMILES O.O.O.O.O.[O-]S(=O)(=S)[O-].[Na+].[Na+]
IUPAC Name disodium;dioxido-oxo-sulfanylidene-$l^{6}-sulfane;pentahydrate
Molecular Weight (g/mol) 248.172
PubChem CID 61475
Formula Weight 158.11
Chemical Name or Material Sodium thiosulfate concentrate, For 1L standard solution, 0.1 M Na2S2O3 (0.1N)
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.