missing translation for 'onlineSavingsMsg'
Learn More

Sodium thiosulfate concentrate, For 1L standard solution, 0.1 M Na2S2O3 (0.1N), Honeywell Fluka™

Product Code. 15645240 Shop All Honeywell Fluka Products
Click to view available options
Quantity:
1 Pc.
6 x 1 Ea.
Unit Size:
Each
Pack of 6
This item is not returnable. View return policy

Product Code. 15645240

Brand: Honeywell-Fluka 382001EA

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

for 1L standard solution, 0.1 M Na2S2O3 (0.1N)

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10102-17-7, 6132-02-1, 7732-18-5
MDL Number MFCD00003499

Specifications

Quantity 1 Pc.
Molecular Formula Na2O3S2
Linear Formula Na2S2O3
UN Number NONH for all modes of transport
Synonym sodium thiosulfate, sodium thiosulphate, disodium thiosulfate, sodium thiosulfate anhydrous, hypo, sodiumthiosulfate, chlorine cure, chlorine control, declor-it, thiosulfuric acid, disodium salt
InChI Key AKHNMLFCWUSKQB-UHFFFAOYSA-L
SMILES [Na+].[Na+].[O-]S([S-])(=O)=O
IUPAC Name disodium;dioxido-oxo-sulfanylidene-$l^{6}-sulfane
Molecular Weight (g/mol) 158.10
PubChem CID 24477
Formula Weight 158.11g/mol
Packaging Ampoule
Chemical Name or Material Sodium thiosulfate concentrate
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.