missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Thiosulfate, Anhydrous, Granular, BAKER ANALYZED™ Reagent, J.T. Baker™
High quality chemicals for laboratory and specialized industrial use
Chemical Identifiers
CAS | 7772-98-7 |
---|---|
Molecular Formula | Na2O3S2 |
Molecular Weight (g/mol) | 158.10 |
MDL Number | MFCD00003499 |
InChI Key | AKHNMLFCWUSKQB-UHFFFAOYSA-L |
Synonym | sodium thiosulfate, sodium thiosulphate, disodium thiosulfate, sodium thiosulfate anhydrous, hypo, sodiumthiosulfate, chlorine cure, chlorine control, declor-it, thiosulfuric acid, disodium salt |
PubChem CID | 24477 |
SMILES | [Na+].[Na+].[O-]S([S-])(=O)=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
15538194
|
Avantor J.T.Baker
3953-01 |
500 g | Poly Bottle |
N/A
|
N/A | ||||
Chemical Identifiers
7772-98-7 | |
158.10 | |
AKHNMLFCWUSKQB-UHFFFAOYSA-L | |
24477 |
Na2O3S2 | |
MFCD00003499 | |
sodium thiosulfate, sodium thiosulphate, disodium thiosulfate, sodium thiosulfate anhydrous, hypo, sodiumthiosulfate, chlorine cure, chlorine control, declor-it, thiosulfuric acid, disodium salt | |
[Na+].[Na+].[O-]S([S-])(=O)=O |
Specifications
7772-98-7 | |
Na2O3S2 | |
sodium thiosulfate, sodium thiosulphate, disodium thiosulfate, sodium thiosulfate anhydrous, hypo, sodiumthiosulfate, chlorine cure, chlorine control, declor-it, thiosulfuric acid, disodium salt | |
[Na+].[Na+].[O-]S([S-])(=O)=O | |
158.10 | |
158.11 | |
1L = 1.67kg |
Granules | |
MFCD00003499 | |
AKHNMLFCWUSKQB-UHFFFAOYSA-L | |
disodium;dioxido-oxo-sulfanylidene-$l^{6}-sulfane | |
24477 | |
BAKER ANALYZED™ Reagent | |
Sodium Thiosulfate, Anhydrous |