missing translation for 'onlineSavingsMsg'
Learn More

Sodium thiosulfate, 98.5%, extra pure, anhydrous

Product Code. 10072510 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
500 g
1 kg
2.5 kg
10 kg
Packaging:
Plastic bottle
Plastic drum
Unit Size:
10kg
1kg
2.5kg
500g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10072510 1 kg Plastic bottle 1kg
10092910 500 g Plastic bottle 500g
10472011 2.5 kg Plastic bottle 2.5kg
11331790 10 kg Plastic drum 10kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10072510 Supplier Thermo Scientific Acros Supplier No. 202870010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7772-98-7 | Na2O3S2 | 158.10 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Reduction reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7772-98-7
Molecular Formula Na2O3S2
Molecular Weight (g/mol) 158.10
MDL Number MFCD00003499
InChI Key AKHNMLFCWUSKQB-UHFFFAOYSA-L
Synonym sodium thiosulfate, sodium thiosulphate, disodium thiosulfate, sodium thiosulfate anhydrous, hypo, sodiumthiosulfate, chlorine cure, chlorine control, declor-it, thiosulfuric acid, disodium salt
PubChem CID 24477
SMILES [Na+].[Na+].[O-]S([S-])(=O)=O

Specifications

Melting Point 48.0°C
Color White
pH 6.0 to 8.5 (5% soln. at 20°C)
Physical Form Crystalline Powder
Quantity 1 kg
Assay Percent Range 98% min. (on dry substance) (Iodimetry)
Linear Formula Na2S2O3
Merck Index 15,8821
Solubility Information Solubility in water: 700g/L (20°C). Other solubilities: practically insoluble in alcohol
IUPAC Name disodium;dioxido-oxo-sulfanylidene-$l^{6}-sulfane
Formula Weight 158.1
Percent Purity 98.5%
Grade Extra Pure
Loss on Drying 2% (105°C, 2 hrs) max.
Packaging Plastic bottle
Chemical Name or Material Sodium thiosulfate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.