missing translation for 'onlineSavingsMsg'
Learn More

Sodium thiosulfate, 0.01N Standardized Solution

Product Code. 11353777
Change view
Click to view available options
Quantity:
1 L
4 L
20 L
Unit Size:
1 L
20 L
4 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11353777 4 L 4 L
11363777 1 L 1 L
11343777 20 L 20 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11353777 Supplier Thermo Scientific Alfa Aesar Supplier No. 035643.K7
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7772-98-7 | Na2O3S2 | 158.10 g/mol

Sodium thiosulfate, 0.01N standardized solution is used in volumetric analysis to estimate the concentration of certain compounds in solution (hydrogen peroxide) as well as chlorine content in bleaching powder and water. It is used as a titrant in iodometry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7772-98-7
Molecular Formula Na2O3S2
Molecular Weight (g/mol) 158.10
MDL Number MFCD00003499
InChI Key AKHNMLFCWUSKQB-UHFFFAOYSA-L
Synonym sodium thiosulfate, sodium thiosulphate, disodium thiosulfate, sodium thiosulfate anhydrous, hypo, sodiumthiosulfate, chlorine cure, chlorine control, declor-it, thiosulfuric acid, disodium salt
PubChem CID 24477
IUPAC Name disodium;dioxido-oxo-sulfanylidene-$l^{6}-sulfane
SMILES [Na+].[Na+].[O-]S([S-])(=O)=O

Specifications

Linear Formula Na2S2O3
Concentration 0.01N
Solubility Information Immiscible with alcohol.
Formula Weight 158.1
Quantity 4 L
Physical Form Liquid
Chemical Name or Material Sodium thiosulfate, Standardized Solution

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.