missing translation for 'onlineSavingsMsg'
Learn More

MuRF1/TRIM63 Antibody (6H6D4) - BSA Free, R&D Systems™

Product Code. 18613918 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
Unit Size:
0.03mg
0.1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18613918 0.025 mg 0.03mg
18275010 0.1 mg 0.1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18613918 Supplier Novus Biologicals Supplier No. NBP2525150.025mg
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Novus product, part of R&D Systems. Mouse Monoclonal Antibody

MuRF1/TRIM63 Monoclonal specifically detects MuRF1/TRIM63 in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MuRF1/TRIM63
Applications ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 6H6D4
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution ELISA 1:10000, Immunohistochemistry 1:200-1:1000, Immunohistochemistry-Paraffin 1:200-1:1000
Formulation PBS with 0.05% Sodium Azide
Gene Alias EC 6.3.2.-, IRFMURF2, Iris RING finger protein, MuRF1, MuRF-1, muscle specific ring finger protein 2, Muscle-specific RING finger protein 1, RING finger protein 28FLJ32380, RNF28E3 ubiquitin-protein ligase TRIM63, SMRZMURF-1, Striated muscle RING zinc finger protein, tripartite motif containing 63, tripartite motif-containing 63, Tripartite motif-containing protein 63
Gene Symbols TRIM63
Host Species Mouse
Immunogen Synthesized peptide of human MuRF1 (AA: 293-304).
Molecular Weight of Antigen 40.2 kDa
Purification Method Protein A or G purified
Quantity 0.025 mg
Regulatory Status RUO
Research Discipline Zinc Finger
Primary or Secondary Primary
Gene ID (Entrez) 84676
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles.
Product Type Antibody
Form Purified
Isotype IgG1
Product Line Novus
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.