missing translation for 'onlineSavingsMsg'
Learn More

Sodium thiomethoxide, 95%, pure, Thermo Scientific Chemicals

Product Code. 10217223
Click to view available options
Quantity:
1 g
5 g
25 g
100 g
Packaging:
Glass bottle

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5188-07-8
Molecular Formula CH3NaS
Molecular Weight (g/mol) 70.09
MDL Number MFCD00174316
InChI Key RMBAVIFYHOYIFM-UHFFFAOYSA-M
PubChem CID 4378561
SMILES C[S-].[Na+]

Specifications

Color White
Infrared Spectrum Authentic
Assay Percent Range 94 to 106% (Iodimetry)
Packaging Glass bottle
Linear Formula CH3SNa
Quantity 25 g
Synonym diethyl 1-tetradecanephosphonate, diethyl 1-tetradecylphosphonate, diethyl tetradecylphosphonate, acmc-20al9h, diethyl tetradecane-1-phosphonate, tetradecylphosphonic acid diethyl ester, tetradecyl-phosphonic acid diethyl ester, 1-tetradecanephosphonic acid diethyl ester, phosphonic acid, tetradecyl-, diethyl ester
Solubility Information Solubility in water: freely soluble.
IUPAC Name sodium methylsulfanide
Formula Weight 70.09
Percent Purity 95%
Grade Pure
Physical Form Powder
Chemical Name or Material Sodium thiomethoxide
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Sodium thiomethoxide
Signal Word
  • Danger
Hazard Category
  • Flammable solid Category 1
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 A
Hazard Statement
  • H228-Flammable solid.
  • H301-Toxic if swallowed.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'