missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Thiocyanate, TRC
Specifications
Specifications
| CAS | 540-72-7 |
| Molecular Formula | C N S . Na |
| Quantity | 25 g |
| Synonym | Thiocyanic acid, sodium salt (1:1), Sodium isothiocyanate (7CI), Thiocyanic acid, sodium salt (8CI,9CI), Haimased, Scyan, Sodium rhodanate, Sodium rhodanide, Sodium sulfocyanate, Sodium sulfocyanide, Sodium thiocyanate, Sodium thiocyanide |
| InChI Formula | InChI=1S/CHNS.Na/c2-1-3;/h3H;/q;+1/p-1 |
| SMILES | [Na+].[S-]C#N |
| IUPAC Name | sodium;thiocyanate |
| Molecular Weight (g/mol) | 81.07 |
| Formula Weight | 80.9649 |
| Chemical Name or Material | Sodium Thiocyanate |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?