missing translation for 'onlineSavingsMsg'
Learn More

Sodium thiocyanate, 98+%, ACS reagent

Product Code. 10229370 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
2.5 kg
Packaging:
Glass bottle
Plastic bottle
Unit Size:
100g
2.5kg
500g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10229370 5 g Glass bottle 5g
10063923 100 g Plastic bottle 100g
10667221 500 g Plastic bottle 500g
10228193 2.5 kg Plastic bottle 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10229370 Supplier Thermo Scientific Acros Supplier No. 419670050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 540-72-7 | CNNaS | 81.07 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 540-72-7
Molecular Formula CNNaS
Molecular Weight (g/mol) 81.07
MDL Number MFCD00011123
InChI Key VGTPCRGMBIAPIM-UHFFFAOYSA-M
Synonym sodium thiocyanate, sodium rhodanate, sodium sulfocyanate, sodium rhodanide, thiocyanic acid, sodium salt, thiocyanate sodium, scyan, sodium isothiocyanate, nascn, haimased
PubChem CID 516871
ChEBI CHEBI:30952
SMILES [Na+].[S-]C#N

Specifications

Melting Point 287.0°C
Color White
Physical Form Crystalline Powder, Crystals and/or Chunks
Quantity 5 g
Assay Percent Range 98% min. (Argentometry)
Linear Formula NaSCN
Fieser 01,1105
Merck Index 15, 9480
Solubility Information Solubility in water: 1390g/L (21°C). Other solubilities: soluble in alcohol, freely soluble in acetone
IUPAC Name sodium;thiocyanate
Formula Weight 81.07
Percent Purity ≥98%
Grade ACS Reagent
Packaging Glass bottle
Chemical Name or Material Sodium thiocyanate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.