missing translation for 'onlineSavingsMsg'
Learn More

Sodium Thiocyanate, ≥98.0% (on dried basis, AT), Honeywell™

Product Code. p-7120836 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
50 g
250 g
1 kg
Packaging:
Poly bottle
Unit Size:
1kg
250g
50g
Product Code. Quantity Packaging unitSize
15636870 50 g Poly bottle 50g
15626870 250 g Poly bottle 250g
15616870 1 kg Poly bottle 1kg
3 options
This item is not returnable. View return policy

Product Code. 15636870

Brand: Honeywell Research Chemicals 7193850G

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Purum p.a.,≥98.0% (on dried basis, AT)

  • Products with a guarantee certificate and/or suitable for the stated analytical application.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 540-72-7
Molecular Formula CNNaS
Molecular Weight (g/mol) 81.07
MDL Number MFCD00011123
InChI Key VGTPCRGMBIAPIM-UHFFFAOYSA-M
Synonym sodium thiocyanate, sodium rhodanate, sodium sulfocyanate, sodium rhodanide, thiocyanic acid, sodium salt, thiocyanate sodium, scyan, sodium isothiocyanate, nascn, haimased
PubChem CID 516871
ChEBI CHEBI:30952
SMILES [Na+].[S-]C#N

Specifications

Quantity 50 g
Linear Formula NaSCN
UN Number NONH for all modes of transport
Beilstein 3594965
IUPAC Name sodium;thiocyanate
Formula Weight 81.07g/mol
Percent Purity ≥98.0% (on dried basis, AT)
Packaging Poly bottle
Chemical Name or Material Sodium thiocyanate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.