missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Sodium tetrathionate dihydrate

Product Code. 15793487 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15793487 100 g 100g
15783487 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15793487 Supplier Thermo Scientific Chemicals Supplier No. 461921000
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 13721-29-4 ∣ H4Na2O8S4 ∣ 306.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13721-29-4
Molecular Formula H4Na2O8S4
Molecular Weight (g/mol) 306.24
MDL Number MFCD00150868
InChI Key HAEPBEMBOAIUPN-UHFFFAOYSA-L
Synonym sodium tetrathionate dihydrate, tetrathionic acid, disodium salt, dihydrate, tetrathionate sodium dihydrate, disodium tetrathionate dihydrate, sodium tetrathionate, dihydrate 6ci,7ci, disodium dihydrate tetrathionate, di dithioperoxo sulfuricacid, sodium salt, hydrate 1:2:2
PubChem CID 203055
IUPAC Name disodium dihydrate (sulfonatodisulfanyl)sulfonate
SMILES O.O.[Na+].[Na+].[O-]S(=O)(=O)SSS([O-])(=O)=O

Specifications

Physical Form Crystalline Powder and/or Crystals
Quantity 100 g
Packaging Glass Bottle
Solubility Information Solubility in water: 30.6g/L.
Formula Weight 306.27
Percent Purity 98%
Grade Ungraded
Chemical Name or Material Sodium tetrathionate dihydrate
compliance-icons
Product Identifier
  • Tetrathionic acid, disodium salt, dihydrate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: Tetrathionic acid, disodium salt, dihydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.