missing translation for 'onlineSavingsMsg'
Learn More

Sodium Tetraphenylboroate, TRC

Product Code. 30415129
Change view
Click to view available options
Quantity:
1 g
250 g
500 g
Unit Size:
250g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30415129 1 g -
30420152 500 g -
30423588 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30415129 Supplier Toronto Research Chemicals Supplier No. TRCS6731001G

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.
, 02 950 59 478

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 143-66-8
Molecular Formula C24 H20 B . Na
Molecular Weight (g/mol) 342.22
Synonym Borate(1-), tetraphenyl-, sodium (1:1) (ACI), Sodium tetraphenylboron, Borate(1-), tetraphenyl-, sodium (8CI, 9CI), HOK-Boron Na, Kalibor, Kalignost, Kariporu K, Sodium tetrakis(phenyl) borate, Sodium tetraphenylborate (NaPh4B), Sodium tetraphenylborate(1-), Sodium tetraphenylboride, Sodium tetraphenylboride(1-), Tetraphenyl boron sodium salt, Tetraphenyl sodium borate
IUPAC Name sodium;tetraphenylboranuide
SMILES [Na+].c1ccc(cc1)[B-](c2ccccc2)(c3ccccc3)c4ccccc4

Specifications

Quantity 1 g
InChI Formula InChI=1S/C24H20B.Na/c1-5-13-21(14-6-1)25(22-15-7-2-8-16-22,23-17-9-3-10-18-23)24-19-11-4-12-20-24;/h1-20H;/q-1;+1
Formula Weight 342.1556
Chemical Name or Material Sodium Tetraphenylboroate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.