missing translation for 'onlineSavingsMsg'
Learn More

Sodium tetrakis(4-fluorophenyl)borate dihydrate, 98%

Product Code. 15412376
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15412376 250 mg 250mg
15492366 1 g 1g
15402376 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15412376 Supplier Thermo Scientific Alfa Aesar Supplier No. H55952.MD
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 207683-22-5 | C24H20BF4NaO2 | 450.215 g/mol

Sodium tetrakis(4-fluorophenyl)borate dihydrate, is used as a reagent for the two-phase titration of nonionic surfactants in the presence of anionic surfactants.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium tetrakis(4-fluorophenyl)borate dihydrate, is used as a reagent for the two-phase titration of nonionic surfactants in the presence of anionic surfactants.

Notes
Hygroscopic. Incompatible withstrong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 207683-22-5
Molecular Formula C24H20BF4NaO2
Molecular Weight (g/mol) 450.215
MDL Number MFCD00149598
InChI Key MSDGDEJOIBMWJD-UHFFFAOYSA-N
Synonym sodium tetrakis 4-fluorophenyl borate dihydrate, cesibor, tetrakis 4-fluorophenyl boron sodium, sodium tetrakis 4-fluorophenyl boranuide dihydrate, sodium tetrakis 4-fluorophenyl borate dihydrate nt, sodium tetrakis 4-fluorophenyl borate dihydrate, selectophore tm
PubChem CID 45073643
IUPAC Name sodium;tetrakis(4-fluorophenyl)boranuide;dihydrate
SMILES [B-](C1=CC=C(C=C1)F)(C2=CC=C(C=C2)F)(C3=CC=C(C=C3)F)C4=CC=C(C=C4)F.O.O.[Na+]

Specifications

Quantity 250 mg
Beilstein 4285808
Formula Weight 450.21 (414.18 Anhydrous)
Percent Purity 98%
Chemical Name or Material Sodium tetrakis(4-fluorophenyl)borate dihydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.