missing translation for 'onlineSavingsMsg'
Learn More

Sodium tetrafluoroborate, 97+%, extra pure

Product Code. 10457940 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
500 g
2.5 kg
Packaging:
Plastic bottle
Unit Size:
2.5kg
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10457940 25 g Plastic bottle 25g
10062610 500 g Plastic bottle 500g
10165103 2.5 kg Plastic bottle 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10457940 Supplier Thermo Scientific Acros Supplier No. 192120250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 13755-29-8 | BF4Na | 109.79 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

C-X bond formation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13755-29-8
Molecular Formula BF4Na
Molecular Weight (g/mol) 109.79
MDL Number MFCD00003515
InChI Key KGJZTOFHXCFQIV-UHFFFAOYSA-N
Synonym sodium tetrafluoroborate, sodium fluoborate, sodium fluoroborate, sodium borofluoride, sodium boron fluoride, apreton r, sodium boron tetrafluoride, sodium fluoroborate nabf4, sodium tetrafluoroborate 1-, sodium tetrafluroborate
PubChem CID 4343483
SMILES [Na+].F[B-](F)(F)F

Specifications

Melting Point 384°C
Color Brown to White
Physical Form Powder, Needles or Crystals
Quantity 25 g
Assay Percent Range 97+%
Linear Formula NaBF4
Merck Index 15, 8753
Solubility Information Solubility in water: 973g/L (20°C). Other solubilities: sparingly soluble in alcohol
IUPAC Name sodium tetrafluoroboranuide
Formula Weight 109.79
Percent Purity ≥97%
Grade Extra Pure
Packaging Plastic bottle
Density 2.47
Chemical Name or Material Sodium tetrafluoroborate, 98%
Show More Show Less
compliance-icons
Product Identifier
  • Sodium tetrafluoroborate
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.