Learn More
Sodium tetrafluoroborate, 97%
Description
Sodium tetrafluoroborate is used in fluxes, metal finishing and in plating circuits. It is used as a starting material in the preparation of ionic liquids as well as boron trifluoride. It acts as an efficient catalyst for the synthesis of 3,4-dihydropyrimidin-2(1H)-ones and thiones.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Sodium tetrafluoroborate is used in fluxes, metal finishing and in plating circuits. It is used as a starting material in the preparation of ionic liquids as well as boron trifluoride. It acts as an efficient catalyst for the synthesis of 3,4-dihydropyrimidin-2(1H)-ones and thiones.
Solubility
Soluble in water.
Notes
Hygroscopic and moisture sensitive. Incompatible with strong oxidizing agents.
Specifications
Specifications
| Melting Point | 384°C (dec.) |
| Quantity | 2 kg |
| Linear Formula | NaBF4 |
| Merck Index | 14,8617 |
| Solubility Information | Soluble in water. |
| Formula Weight | 109.79 |
| Percent Purity | 97% |
| Sensitivity | Hygroscopic |
| Density | 2.47 |
| Chemical Name or Material | Sodium tetrafluoroborate |
Safety and Handling
- Danger
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.