missing translation for 'onlineSavingsMsg'
Learn More

Sodium tetraethylborate, 98%

Product Code. 11389389
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11389389 1 g 1g
11379389 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11389389 Supplier Thermo Scientific Alfa Aesar Supplier No. 089137.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 15523-24-7 | C8H20BNa | 150.048 g/mol

Sodium tetraethylborate is used as a derivatizing agent for the detection of organotin, lead and mercury compounds by using gas chromatography coupled with various sensitive and selective detection methods, such as atomic absorption spectrometry (AAS), atomic emission spectrometry (AES), pulsed-flame photometric detection (PFPD), and mass spectrometry (MS). It is also used as an alkylating agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15523-24-7
Molecular Formula C8H20BNa
Molecular Weight (g/mol) 150.048
MDL Number MFCD00061547
InChI Key SZSBMTRYJRHYNI-UHFFFAOYSA-N
Synonym sodium tetraethylborate, sodium tetraethylboranuide, borate 1-,tetraethyl-, sodium 1:1, sodium tetraethylborate 1-, et4bna, sodiotetraethylboron v, borate 1-, tetraethyl-, sodium, sodium tetraethylborate 1g
PubChem CID 23681030
IUPAC Name sodium;tetraethylboranuide
SMILES [B-](CC)(CC)(CC)CC.[Na+]

Specifications

Melting Point (decomposition)
Assay Percent Range 98%
Linear Formula NaB(C2H5)4
Quantity 1 g
UN Number UN3393
Sensitivity Air and moisture sensitive
Solubility Information Dissolves in water with decomposition
Formula Weight 150.05
Percent Purity 98%
Physical Form Crystalline
Chemical Name or Material Sodium tetraethylborate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.