missing translation for 'onlineSavingsMsg'
Learn More

Sodium tetracyanoplatinate(II) hydrate, Premion™, 99.95% (metals basis), Pt 48.4% min, Thermo Scientific Chemicals

Product Code. 11393988
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11393988 1 g 1g
11303998 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11393988 Supplier Thermo Scientific Alfa Aesar Supplier No. 040400.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 699012-94-7
Molecular Formula C4N4Na2Pt
Molecular Weight (g/mol) 345.13
MDL Number MFCD00798527
InChI Key WOIFBVMIVVRIIE-UHFFFAOYSA-N
Synonym disodium tetracyanoplatinumdiuide hydrate, sodium tetracyanoplatinate ii hydrate, premion
PubChem CID 16217382
IUPAC Name disodium tetracyanoplatinumbis(ylium)
SMILES [Na+].[Na+].N#C[Pt++](C#N)(C#N)C#N

Specifications

Melting Point (decomposition)
Color White
Physical Form Crystalline Powder
Quantity 1 g
Formula Weight 345.14 (Anhydrous)
Percent Purity 99.95%
Grade Premion
Concentration or Composition (by Analyte or Components) Pt 48.4% min.
Assay (metals basis)
Chemical Name or Material Sodium tetracyanoplatinate(II) hydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.