missing translation for 'onlineSavingsMsg'
Learn More

Sodium tetrachloropalladate(II) hydrate, Premion™, 99.999% (metals basis), Pd 30% min

Product Code. 11384988
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11384988 1 g 1g
11374988 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11384988 Supplier Thermo Scientific Alfa Aesar Supplier No. 043945.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 13820-53-6 | Cl4Na2Pd | 294.20 g/mol

Sodium tetrachloropalladate is used in organ metallic chemistry for the preparation of phosphine complexes of palladium.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13820-53-6
Molecular Formula Cl4Na2Pd
Molecular Weight (g/mol) 294.20
MDL Number MFCD00003487
InChI Key ABKQFSYGIHQQLS-UHFFFAOYSA-J
Synonym sodium tetrachloropalladate ii, disodium tetrachloropalladate, palladium ii sodium chloride, sodiumpalladiumchloride, disodiumpalladiumtetrachloride, sodiumtetrachloropalladate ii, palladiumsodiumchloridetrihydrate, disodiumtetrachloropalladate 2-, tetrachloro-palladate 2-disodium, disodium;tetrachloropalladium 2-
PubChem CID 11000870
IUPAC Name disodium;tetrachloropalladium(2-)
SMILES [Na+].[Na+].Cl[Pd++](Cl)(Cl)Cl

Specifications

Color Red to Brown
Physical Form Crystalline
Quantity 1 g
Linear Formula Na2PdCl4·xH2O (x≈3)
Solubility Information Soluble in water and ethyl alcohol.
Formula Weight 294.20 (Anhydrous)
Percent Purity 99.999%
Grade Premion
Concentration or Composition (by Analyte or Components) Pd 30% min
Sensitivity Hygroscopic
Assay (metals basis)
Chemical Name or Material Sodium tetrachloropalladate (II) hydrate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.