Learn More
Sodium tetrachloropalladate(II) hydrate, 99.95% (metals basis), Pd 30%
Descripción
Sodium tetrachloropalladate is used as a test salt for Pd allergy patch testing. It is used in chemical synthesis as a catalyst.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Especificaciones
Especificaciones
| Color | Red to Brown |
| Linear Formula | Na2PdCl4·xH2O (x≈3) |
| Quantity | 2 g |
| Sensitivity | Hygroscopic |
| Solubility Information | Soluble in water and acetic acid. |
| Formula Weight | 294.20 (Anhydrous) |
| Percent Purity | 99.95% |
| Concentration or Composition (by Analyte or Components) | Pd 30% |
| Physical Form | Crystalline |
| Assay | (metals basis) |
| Mostrar más |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.