missing translation for 'onlineSavingsMsg'
Learn More

Sodium tetrachloropalladate(II) hydrate, 99.95% (metals basis), Pd 30%

Product Code. 11321978
Click to view available options
Quantity:
2 g
10 g
50 g
Unit Size:
10g
2g
50g
This item is not returnable. View return policy

Product Code. 11321978

Brand: Thermo Scientific Alfa Aesar 011886.09

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 13820-53-6 | Cl4Na2Pd | 294.20 g/mol

Sodium tetrachloropalladate is used as a test salt for Pd allergy patch testing. It is used in chemical synthesis as a catalyst.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13820-53-6
Molecular Formula Cl4Na2Pd
Molecular Weight (g/mol) 294.20
MDL Number MFCD00003487
InChI Key ABKQFSYGIHQQLS-UHFFFAOYSA-J
Synonym sodium tetrachloropalladate ii, disodium tetrachloropalladate, palladium ii sodium chloride, sodiumpalladiumchloride, disodiumpalladiumtetrachloride, sodiumtetrachloropalladate ii, palladiumsodiumchloridetrihydrate, disodiumtetrachloropalladate 2-, tetrachloro-palladate 2-disodium, disodium;tetrachloropalladium 2-
PubChem CID 11000870
IUPAC Name disodium;tetrachloropalladium(2-)
SMILES [Na+].[Na+].Cl[Pd++](Cl)(Cl)Cl

Specifications

Color Red to Brown
Linear Formula Na2PdCl4·xH2O (x≈3)
Quantity 10 g
Sensitivity Hygroscopic
Solubility Information Soluble in water and acetic acid.
Formula Weight 294.20 (Anhydrous)
Percent Purity 99.95%
Concentration or Composition (by Analyte or Components) Pd 30%
Physical Form Crystalline
Assay (metals basis)
Chemical Name or Material Sodium tetrachloropalladate(II) hydrate
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.