missing translation for 'onlineSavingsMsg'
Learn More

Sodium tetrachloropalladate(II), approx. 36% Pd, Thermo Scientific Chemicals

Catalog Number 10241832
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
13820-53-6
Cl4Na2Pd
294.20
MFCD00003487
ABKQFSYGIHQQLS-UHFFFAOYSA-J
sodium tetrachloropalladate ii, disodium tetrachloropalladate, palladium ii sodium chloride, sodiumpalladiumchloride, disodiumpalladiumtetrachloride, sodiumtetrachloropalladate ii, palladiumsodiumchloridetrihydrate, disodiumtetrachloropalladate 2-, tetrachloro-palladate 2-disodium, disodium;tetrachloropalladium 2-
11000870
[Na+].[Na+].Cl[Pd++](Cl)(Cl)Cl
This item is not returnable. View return policy
13820-53-6
Cl4Na2Pd
294.20
MFCD00003487
ABKQFSYGIHQQLS-UHFFFAOYSA-J
sodium tetrachloropalladate ii, disodium tetrachloropalladate, palladium ii sodium chloride, sodiumpalladiumchloride, disodiumpalladiumtetrachloride, sodiumtetrachloropalladate ii, palladiumsodiumchloridetrihydrate, disodiumtetrachloropalladate 2-, tetrachloro-palladate 2-disodium, disodium;tetrachloropalladium 2-
11000870
[Na+].[Na+].Cl[Pd++](Cl)(Cl)Cl

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Brown to Orange or Brown-Green
Physical Form Powder and Granules, Crystals
Quantity 1 g
Assay Percent Range 36%
Linear Formula Na2PdCl4
Fieser 05,626; 06,45; 09,207; 16,309; 01,778; 04,365; 05,498; 06,443; 07,274; 15,13; 16,356
Solubility Information Solubility in water: soluble
IUPAC Name disodium;tetrachloropalladium(2-)
Formula Weight 294.21
Percent Purity ∼36% Pd
Packaging Glass bottle
Chemical Name or Material Sodium tetrachloropalladate(II), ca. 36.4% Pd
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Sodium tetrachloropalladate(II)
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Substances and mixtures corrosive to metals Category 1
Hazard Statement
  • H319-Causes serious eye irritation.
  • H302-Harmful if swallowed.
  • H290-May be corrosive to metals.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'