missing translation for 'onlineSavingsMsg'
Learn More

Sodium tetrachloroaurate(III) dihydrate, Premion™, 99.99% (metals basis), Au 49-50%

Product Code. 11376518
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11376518 5 g 5g
11366518 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11376518 Supplier Thermo Scientific Alfa Aesar Supplier No. 012148.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

CAS: 13874-02-7 | AuCl4H4NaO2 | 397.79 g/mol

It is used in chemo-therapeutics. It is employed as a catalyst in the removal of tert-butyl(dimethyl)silyl (TBS) protecting groups. It is selective in the presence of aromatic TBS ethers, aliphatic triisopropylsilyl ethers, aliphatic tert-butyl(diphenyl)silyl ethers, or sterically hindered aliphatic TBS ethers. With excess catalyst, TBS ethers can also be transformed into 4-methoxybenzyl ethers or methyl ethers in one pot.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13874-02-7
Molecular Formula AuCl4H4NaO2
Molecular Weight (g/mol) 397.79
MDL Number MFCD00149162
InChI Key OEERDTKOVZFOHF-UHFFFAOYSA-J
Synonym sodium tetrachloroaurate iii dihydrate, sodium;tetrachlorogold 1-;dihydrate, sodiumtetrachloroaurate iii dihydrate, sodium dihydrate tetrachloroaurate
PubChem CID 18949434
SMILES O.O.[Na+].Cl[Au-](Cl)(Cl)Cl

Specifications

Melting Point 100°C (decomposition)
Physical Form Crystalline
Quantity 5 g
Assay Percent Range 99.99% (metals basis)
Solubility Information Soluble in water,alcohol and ether.
IUPAC Name sodium;tetrachlorogold(1-);dihydrate
Formula Weight 397.80 (361.77 Anhydrous)
Density 0.8 g/mL
Chemical Name or Material Sodium tetrachloroaurate(III) dihydrate
compliance-icons compliance-icons
Product Identifier
  • Sodium tetrachloroaurate(III) dihydrate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 1
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H290-May be corrosive to metals.
  • H302-Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: Sodium tetrachloroaurate(III) dihydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.