missing translation for 'onlineSavingsMsg'
Learn More

Sodium tetrabromoaurate(III) hydrate, 99.99% (Metals basis), Thermo Scientific Chemicals, Premion™

Product Code. p-4782155
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11334028

Brand: Thermo Scientific Alfa Aesar 040428.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Sodium tetrabromoaurate(III) hydrate participates in the fabricated of core/alloy nanoparticles via the layer-by-layer hydrothermal route ans its growth characteristics and optical properties study.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10378-49-1
Molecular Formula AuBr4Na
Molecular Weight (g/mol) 539.57
MDL Number MFCD00274624
InChI Key PMPPMRNISYJZFW-UHFFFAOYSA-J
Synonym sodium tetrabromoaurate iii hydrate, aubr4.na.h2o, gold sodium bromide, naaubr4*xh2o, sodium tetrabromoaurate iii hydrate, premion, sodium tetrabromoaurate 1--water 1/1/1
PubChem CID 71310037
SMILES [Na+].Br[Au-](Br)(Br)Br

Specifications

Physical Form Crystalline
Quantity 1 g
Assay Percent Range 99.99% (metals basis)
Solubility Information Soluble in water
IUPAC Name sodium;tetrabromogold(1-);hydrate
Formula Weight 539.59 (Anhydrous)
Odor Odorless
Appearance Crystalline powder
Chemical Name or Material Sodium tetrabromoaurate(III) hydrate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.