missing translation for 'onlineSavingsMsg'
Learn More

Sodium tetrabromoaurate(III) hydrate, 99.99% (Metals basis), Thermo Scientific Chemicals, Premion™

Product Code. 11344028
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11344028 5 g 5g
11334028 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11344028 Supplier Thermo Scientific Alfa Aesar Supplier No. 040428.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Sodium tetrabromoaurate(III) hydrate participates in the fabricated of core/alloy nanoparticles via the layer-by-layer hydrothermal route ans its growth characteristics and optical properties study.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10378-49-1
Molecular Formula AuBr4Na
Molecular Weight (g/mol) 539.57
MDL Number MFCD00274624
InChI Key PMPPMRNISYJZFW-UHFFFAOYSA-J
Synonym sodium tetrabromoaurate iii hydrate, aubr4.na.h2o, gold sodium bromide, naaubr4*xh2o, sodium tetrabromoaurate iii hydrate, premion, sodium tetrabromoaurate 1--water 1/1/1
PubChem CID 71310037
SMILES [Na+].Br[Au-](Br)(Br)Br

Specifications

Physical Form Crystalline
Quantity 5 g
Assay Percent Range 99.99% (metals basis)
Solubility Information Soluble in water
IUPAC Name sodium tetrabromogolduide hydrate
Formula Weight 539.59 (Anhydrous)
Odor Odorless
Appearance Crystalline powder
Chemical Name or Material Sodium tetrabromoaurate(III) hydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.