missing translation for 'onlineSavingsMsg'
Learn More

Sodium Tetraborate Decahydrate (White Crystals/Molecular Biology), Fisher BioReagents

Product Code. 10112913
Change view
Click to view available options
Quantity:
500 g
Packaging:
Glass Bottle
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10112913 500 g Glass Bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10112913 Supplier Fisher BioReagents Supplier No. BP175500
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 1303-96-4 | B4Na2O7 · 10 H2O

Glass Bottle; 500g

Sodium Borate can be used in the preparation of TBE electrophoresis buffer.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Sodium Tetraborate Decahydrate
Melting Point 75°C
CAS 1303-96-4, 1330-43-4
Color White
pH 9
Physical Form Powder/Solid
Quantity 500 g
Assay Percent Range 99.5 to 105.0 %
Molecular Formula B4Na2O7 · 10 H2O
Merck Index 15, 8726
Synonym Borax, Boric Acid, Disodium Salt
Formula Weight 381.36
Percent Purity 99.5 to 105.0%
Grade Molecular Biology
Identification Pass Test
Packaging Glass Bottle
Density 1.7300g/cm³
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Sodium tetraborate decahydrate
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Reproductive toxicity Category 1B
Hazard Statement
  • H319-Causes serious eye irritation.
  • H360FD-May damage fertility. May damage the unborn child.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.