missing translation for 'onlineSavingsMsg'
Learn More

Sodium tetraborate decahydrate, Puriss., meets analytical spec. of BP, NF, Ph. Eur., 99.0-103.0%, Solstice
BIOPHARMA

Product Code. 15631070
Change view
Click to view available options
Quantity:
2.5 kg
25 kg
6 x 2.5 kg
Packaging:
Paper / Poly Bag
Poly Bottle
Unit Size:
2.5kg
25kg
Pack of 6
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15631070 6 x 2.5 kg Poly Bottle Pack of 6
15611070 2.5 kg Poly Bottle 2.5kg
15621070 25 kg Paper / Poly Bag 25kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15631070 Supplier Honeywell-Fluka Supplier No. 116256X2.5KG
  • ONLINE EXCLUSIVE PRICE
    £469.00
    £431.63 / Pack of 6
    Save £37.37
    7% Off
 Restricted Item
Estimated Shipment: 22-02-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Puriss., meets analytical specification of BP, NF, Ph. Eur., 99.0-103.0%

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1303-96-4
MDL Number MFCD00149193
Synonym Borax decahydrate, Sodium borate decahydrate

Specifications

Boiling Point 1,575°C
CAS Max % 100.0000%
Quantity 6 x 2.5 kg
Assay Percent Range 99 to 103.0%
Linear Formula Na2B4O7 · 10H2O
UN Number NONH for all modes of transport
Formula Weight 381.37
Percent Purity 99.0 to 103.0%
Grade puriss.
Packaging Poly Bottle
Density 1.73 g/mL at 25 °C(lit.)
Chemical Name or Material Sodium tetraborate decahydrate, Puriss., meets analytical specification of BP, NF, Ph. Eur., 99.0-103.0%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.