missing translation for 'onlineSavingsMsg'
Learn More

Sodium tetraborate decahydrate, 99.5%, for analysis

Product Code. p-3734676 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 kg
5 kg
10 kg
Packaging:
Plastic Bottle
Plastic drum
Unit Size:
10kg
1kg
5kg
This item is not returnable. View return policy

Product Code. 10593371

Brand: Thermo Scientific Acros 205950010

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1303-96-4 | B4Na2O7·10H2O | 381.36 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Biological buffer for analysis
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1303-96-4
Molecular Formula B4Na2O7·10H2O
Molecular Weight (g/mol) 381.36
MDL Number MFCD00149193
Synonym Borax, Sodium borate decahydrate

Specifications

Melting Point 75.0°C
Color White
pH 9 to 9.5 (5% soln. at 20°C)
Physical Form Fine Crystalline Powder
Quantity 1 kg
Assay Percent Range 99.5%
Linear Formula Na2B4O7·10H2O
Fieser 04,461
Merck Index 15,8726
Solubility Information Solubility in water: 60g/L (20°C). Other solubilities: soluble in glycerol, insoluble in alcohol
Formula Weight 381.36
Percent Purity 99.5%
Grade Analytical
Packaging Plastic Bottle
Chemical Name or Material Sodium tetraborate decahydrate
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Sodium tetraborate decahydrate
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Reproductive toxicity Category 1B
Hazard Statement
  • H319-Causes serious eye irritation.
  • H360FD-May damage fertility. May damage the unborn child.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.