missing translation for 'onlineSavingsMsg'
Learn More

Sodium tetraborate, anhydrous, 99.95% (metals basis)

Product Code. 11365049
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11365049 25 g 25g
11355049 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11365049 Supplier Thermo Scientific Alfa Aesar Supplier No. 087862.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1330-43-4 | B4Na2O7 | 201.21 g/mol

Sodium tetraborate is a useful anhydrous borax for proteomics research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1330-43-4
Molecular Formula B4Na2O7
Molecular Weight (g/mol) 201.21
MDL Number MFCD00081185
InChI Key NXSVMNYRVBQCMO-UHFFFAOYSA-N
Synonym Borax
IUPAC Name disodium bicyclo[3.3.1]tetraboroxane-3,7-bis(ylium)-1,5-diuide-1,5-bis(olate)
SMILES [Na+].[Na+].[O-][B-]12O[B+]O[B-]([O-])(O[B+]O1)O2

Specifications

Boiling Point 1575°C (decomposition)
Melting Point 741°C
Color White
Physical Form Powder
Quantity 25 g
Assay Percent Range 99.95% (metals basis)
Solubility Information Slowly soluble in water (87.9g/L).
Formula Weight 201.22
Odor Odorless
Refractive Index 1.501
Sensitivity Hygroscopic
Density 2.637 g/cm3 at 20°C
Chemical Name or Material Sodium tetraborate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.