missing translation for 'onlineSavingsMsg'
Learn More

Sodium tetraborate, anhydrous, 99.5% (metals basis)

Product Code. 11391659
Change view
Click to view available options
Quantity:
1 kg
5 kg
Unit Size:
1kg
5kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11391659 5 kg 5kg
11381659 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11391659 Supplier Thermo Scientific Alfa Aesar Supplier No. 012305.A7
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1330-43-4 | B4Na2O7 | 201.21 g/mol

Sodium tetraborate is used as a co-catalyst for (2,2,6,6-tetramethylpiperidin-1-yl)oxyl and sodium hypochlorite oxidation of alcohols in greener and non-chlorinated solvents. It acts as a reagent for the decomposition of oxides and silicates by fusion. It is used to prepare buffers, which is used in gel electrophoresis of DNA. It acts as a source of borate, micronutrient fertilizer, preservative in taxidermy, additive in ceramic slips and glazes. It is also used as a precursor for sodium perborate monohydrate and boric acid. It finds application as fluoride detoxification, neutron absorber in nuclear reactors and to make indelible ink for dip pens.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1330-43-4
Molecular Formula B4Na2O7
Molecular Weight (g/mol) 201.21
MDL Number MFCD00081185
InChI Key NXSVMNYRVBQCMO-UHFFFAOYSA-N
Synonym Borax, anhydrous, Sodium borate (tetra)
IUPAC Name disodium bicyclo[3.3.1]tetraboroxane-3,7-bis(ylium)-1,5-diuide-1,5-bis(olate)
SMILES [Na+].[Na+].[O-][B-]12O[B+]O[B-]([O-])(O[B+]O1)O2

Specifications

Boiling Point 1575°C (decomposition)
Melting Point 741°C
Color Green
Physical Form Granules
Quantity 5 kg
Assay Percent Range 99.5% (metals basis)
Solubility Information Slowly soluble in waterSoluble in water and glycerol. Slightly soluble in ethanol. Insoluble in acids.
Formula Weight 201.22
Odor Odorless
Refractive Index 1.501
Sensitivity Hygroscopic
Appearance White powder
Vapor Pressure 0 hPa
Density 2.367 g/cm3 at 20°C
Chemical Name or Material Sodium tetraborate, anhydrous
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.