missing translation for 'onlineSavingsMsg'
Learn More

Sodium tetraborate, 98%, for analysis, anhydrous, ACROS Organics™

Brand:  Acros Organics 206295000

Code : 29

Additional Details : CAS Number : 1330-43-4 Weight : 0.50000kg


Packaging Plastic bottle
Quantity 500g

 View more versions of this product

Product Code. 10042780

  • £49.34 / 500g

In stock, ready to ship: 15

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Chemical Identifiers

1330-43-4
B4Na2O7
201.22
Borax, fused

Specifications

Sodium tetraborate
98%
Na2B4O7
15,8726
Solubility in water: 26g/L (20°C). Other solubilities: insoluble in alcohol
201.22
98%
200ppm max.
500ppm max.
50ppm max.
0.05% max. (in water)
50ppm max.
50ppm max.
50ppm max.
Solubility in water: 26g/L (20°C). Other solubilities: insoluble in alcohol
Orange
500g
1330-43-4
B4Na2O7
04,461; 15,295
Borax, fused
201.22
Crystals or Crystalline Powder
Analysis
500ppm max.
50ppm max.
0.05 % max.
50ppm max.
200ppm max.
Plastic bottle
500ppm max.
1575.0°C
741.0°C
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Sodium tetraborate, anhydrous

Signal Word
  • Danger

Hazard Category
  • Reproductive toxicity Category 2

Hazard Statement
  • H360FD-May damage fertility. May damage the unborn child.

Precautionary Statement
  • P201-Obtain special instructions before use.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

SDS Documents
Certificates