missing translation for 'onlineSavingsMsg'
Learn More

Sodium tert-butoxide, 97%

Product Code. 11458797
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
Unit Size:
100g
2500g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11458797 500 g 500g
11448797 100 g 100g
11468797 2500 g 2500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11458797 Supplier Thermo Scientific Alfa Aesar Supplier No. A19942.36
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 865-48-5 | C4H9NaO | 96.105 g/mol

Sodium tert-butoxide is used as a strong base and a non-nucleophilic base. In organic synthesis, it serves as an intermediate in various reactions like condensation, rearrangement and ring-opening. Further, it finds use in agrochemicals, pharmaceuticals, colorants, aroma chemicals, detergents and biodiesel. It acts as a catalyst in polymerization and isomerization reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium tert-butoxide is used as a strong base and a non-nucleophilic base. In organic synthesis, it serves as an intermediate in various reactions like condensation, rearrangement and ring-opening. Further, it finds use in agrochemicals, pharmaceuticals, colorants, aroma chemicals, detergents and biodiesel. It acts as a catalyst in polymerization and isomerization reactions.

Notes
Air sensitive and hygroscopic. Incompatible with acids, reducing agents, oxygen, water, alcohols, chlorinated solvents and halogens.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 865-48-5
Molecular Formula C4H9NaO
Molecular Weight (g/mol) 96.105
MDL Number MFCD00040575
InChI Key MFRIHAYPQRLWNB-UHFFFAOYSA-N
Synonym sodium tert-butoxide, sodium 2-methylpropan-2-olate, sodium t-butoxide, sodium-t-butoxide, t-butoxy sodium, tert-butoxysodium, naotbu, tbuona, 2-propanol, 2-methyl-, sodium salt, sodium-tert-butoxide
PubChem CID 23676156
IUPAC Name sodium;2-methylpropan-2-olate
SMILES CC(C)(C)[O-].[Na+]

Specifications

Density 0.785
Odor Odorless
Linear Formula (CH3)3CONa
Quantity 500 g
UN Number UN3206
Beilstein 3654215
Sensitivity Air and moisture sensitive
Formula Weight 96.11
Percent Purity 97%
Chemical Name or Material Sodium tert-butoxide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.