missing translation for 'onlineSavingsMsg'
Learn More

Sodium Terephthalate, MP Biomedicals

Product Code. 11437315 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
10 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11437315 10 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11437315 Supplier MP Biomedicals Supplier No. 0520943805
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 10028-70-3
Molecular Formula C8H4Na2O4
Quantity 10 g
Synonym disodium terephthalate, terephthalic acid disodium salt, terephthalic acid, disodium salt, sodium terephthalate, unii-gi30zoc0oo, gi30zoc0oo, disodiumterephthalate, acmc-1bvw0, disodium terephthalate 5g, 1,4-benzenedicarboxylicacid, sodium salt 1:?
InChI Key VIQSRHWJEKERKR-UHFFFAOYSA-L
SMILES C1=CC(=CC=C1C(=O)[O-])C(=O)[O-].[Na+].[Na+]
IUPAC Name disodium;terephthalate
Molecular Weight (g/mol) 210.096
PubChem CID 82305
Chemical Name or Material Sodium Terephthalate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.