missing translation for 'onlineSavingsMsg'
Learn More

Sodium tellurite(IV), 99.5% (metals basis)

Product Code. 11398918
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11398918 10 g 10g
11308928 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11398918 Supplier Thermo Scientific Alfa Aesar Supplier No. 041778.09
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 10102-20-2 | H10Na2O9Te | 327.65 g/mol

Sodium tellurite(IV) acts as an intermediate used in the extraction of the tellurium. It is used to make corrosion-resistant electroplated nickel. It is also used to add a bluish coating to iron, steel and aluminum. It serves as a growth medium for bacteria in microbiology.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10102-20-2
Molecular Formula H10Na2O9Te
Molecular Weight (g/mol) 327.65
MDL Number MFCD00036141
InChI Key OVBVNEXDNNHXIU-UHFFFAOYSA-L
Synonym sodium tellurite, disodium tellurite, sodium tellurate iv, disodium trioxotellurate, unii-0bb57la23y, tellurous acid, disodium salt, sodium tellurite iv, telluric acid h2teo3 , disodium salt, telluric acid, disodium salt
PubChem CID 24935
IUPAC Name disodium;tellurite
SMILES O.O.O.O.O.[Na+].[Na+].[O-][Te]([O-])(=O)=O

Specifications

Color White
Physical Form Powder
Quantity 10 g
Assay Percent Range 99.5% (metals basis)
Solubility Information Soluble in water.
Formula Weight 221.58
Appearance White powder
Chemical Name or Material Sodium tellurite(IV)
compliance-icons
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
Hazard Statement
  • H301-Toxic if swallowed.
Precautionary Statement
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
Supplemental information
  • MIXTURE LIST-Contain: Sodium tellurite

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.