missing translation for 'onlineSavingsMsg'
Learn More

Sodium tellurite(IV), 99.5% (metals basis)

Product Code. p-4407942
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
This item is not returnable. View return policy

Product Code. 11398918

Brand: Thermo Scientific Alfa Aesar 041778.09

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 10102-20-2 | H10Na2O9Te | 327.65 g/mol

Sodium tellurite(IV) acts as an intermediate used in the extraction of the tellurium. It is used to make corrosion-resistant electroplated nickel. It is also used to add a bluish coating to iron, steel and aluminum. It serves as a growth medium for bacteria in microbiology.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10102-20-2
Molecular Formula H10Na2O9Te
Molecular Weight (g/mol) 327.65
MDL Number MFCD00036141
InChI Key OVBVNEXDNNHXIU-UHFFFAOYSA-L
Synonym sodium tellurite, disodium tellurite, sodium tellurate iv, disodium trioxotellurate, unii-0bb57la23y, tellurous acid, disodium salt, sodium tellurite iv, telluric acid h2teo3 , disodium salt, telluric acid, disodium salt
PubChem CID 24935
IUPAC Name disodium;tellurite
SMILES O.O.O.O.O.[Na+].[Na+].[O-][Te]([O-])(=O)=O

Specifications

Color White
Physical Form Powder
Quantity 10 g
Assay Percent Range 99.5% (metals basis)
Solubility Information Soluble in water.
Formula Weight 221.58
Appearance White powder
Chemical Name or Material Sodium tellurite(IV)
compliance-icons
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
Hazard Statement
  • H301-Toxic if swallowed.
Precautionary Statement
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
Supplemental information
  • MIXTURE LIST-Contains : Sodium tellurite

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.