missing translation for 'onlineSavingsMsg'
Learn More

Sodium taurocholate hydrate, 96%

Product Code. 11463907
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11463907 25 g 25g
11443907 1 g 1g
11453907 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11463907 Supplier Thermo Scientific Alfa Aesar Supplier No. A18346.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

Sodium taurocholate hydrate (CAS # 145-42-6) is a bile acid and detergent used in molecular biology for protein solubilization. | CAS: 345909-26-4 | C26H44NNaO7S | 537.688 g/mol

Sodium taurocholate is a bile acid having detergent capabilities; natural surfactant. It is used for solubilization of protein. In the preparation of digestive enzymes, it is used as an emulsifier.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • Sodium taurocholate hydrate is a bile acid conjugate of taurine and cholic acid that acts as a detergent
  • Sodium taurocholate solubilizes fats within the digestive system
  • It is the main component of bile acid in carnivorous animals; metabolites are also common in human bile

Application

  • Sodium taurocholate hydrate can be used as an emulsifier for protein isolation
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 345909-26-4
Molecular Formula C26H44NNaO7S
Molecular Weight (g/mol) 537.688
MDL Number MFCD00150819
InChI Key JAJWGJBVLPIOOH-VXFFEJGCSA-M
Synonym sodium taurocholate, taurocholic acid sodium salt, taurocholate sodium, taurocholate sodium salt, unii-m6n3th81no, monosodium n-choloyltaurinate, m6n3th81no, monosodium taurocholate, sodium n-choloyltaurinate, monosodium taurocholic acid
PubChem CID 131632374
IUPAC Name sodium;2-[[(4R)-4-[(3R,5S,7R,8R,9R,10S,12S,13R,14S,17R)-3,7,12-trihydroxy-10,13-dimethyl-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-17-yl]pentanoyl]amino]ethanesulfonate
SMILES CC(CCC(=O)NCCS(=O)(=O)[O-])C1CCC2C1(C(CC3C2C(CC4C3(CCC(C4)O)C)O)O)C.[Na+]

Specifications

Assay Percent Range 0.97
Quantity 25 g
Solubility Information Soluble in Water (50mg/mL- Clear Solution)
Formula Weight 537.73 (Anhydrous)
Chemical Name or Material Sodium Taurocholate Hydrate
compliance-icons
Product Identifier
  • Sodium taurocholate hydrate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P271-Use only outdoors or in a well-ventilated area.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P362+P364-Take off contaminated clothing and wash it before reuse.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: TAUROCHOLIC ACID, SODIUM SALT HYDRATE

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.