missing translation for 'onlineSavingsMsg'
Learn More

Sodium taurocholate hydrate, 96%

Product Code. p-3173441
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 11443907

Brand: Thermo Scientific Alfa Aesar A18346.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

Sodium taurocholate hydrate (CAS # 145-42-6) is a bile acid and detergent used in molecular biology for protein solubilization. | CAS: 345909-26-4 | C26H44NNaO7S | 537.688 g/mol

Sodium taurocholate is a bile acid having detergent capabilities; natural surfactant. It is used for solubilization of protein. In the preparation of digestive enzymes, it is used as an emulsifier.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • Sodium taurocholate hydrate is a bile acid conjugate of taurine and cholic acid that acts as a detergent
  • Sodium taurocholate solubilizes fats within the digestive system
  • It is the main component of bile acid in carnivorous animals; metabolites are also common in human bile

Application

  • Sodium taurocholate hydrate can be used as an emulsifier for protein isolation
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 345909-26-4
Molecular Formula C26H44NNaO7S
Molecular Weight (g/mol) 537.688
MDL Number MFCD00150819
InChI Key JAJWGJBVLPIOOH-VXFFEJGCSA-M
Synonym sodium taurocholate, taurocholic acid sodium salt, taurocholate sodium, taurocholate sodium salt, unii-m6n3th81no, monosodium n-choloyltaurinate, m6n3th81no, monosodium taurocholate, sodium n-choloyltaurinate, monosodium taurocholic acid
PubChem CID 131632374
IUPAC Name sodium;2-[[(4R)-4-[(3R,5S,7R,8R,9R,10S,12S,13R,14S,17R)-3,7,12-trihydroxy-10,13-dimethyl-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-17-yl]pentanoyl]amino]ethanesulfonate
SMILES CC(CCC(=O)NCCS(=O)(=O)[O-])C1CCC2C1(C(CC3C2C(CC4C3(CCC(C4)O)C)O)O)C.[Na+]

Specifications

Assay Percent Range 0.97
Quantity 1 g
Solubility Information Soluble in Water (50mg/mL- Clear Solution)
Formula Weight 537.73 (Anhydrous)
Chemical Name or Material Sodium taurocholate hydrate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.