missing translation for 'onlineSavingsMsg'
Learn More

Sodium Sulphate Decahydrate, TRC

Product Code. 30419854
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
Product Code. Quantity unitSize
30419854 25 g 25g
1 options
This item is not returnable. View return policy

Product Code. 30419854

Brand: Toronto Research Chemicals TRCS22440025G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Sodium Sulphate Decahydrate
CAS 7727-73-3
Molecular Formula 2 Na . O4 S . 10[H2 O]
Quantity 25 g
Synonym Sodium sulfate decahydrate
InChI Formula InChI=1S/2Na.H2O4S.10H2O/c;;1-5(2,3)4;;;;;;;;;;/h;;(H2,1,2,3,4);10*1H2/q2*+1;;;;;;;;;;;/p-2
SMILES O.O.O.O.O.O.O.O.O.O.[Na+].[Na+].[O-]S(=O)(=O)[O-]
IUPAC Name disodium;sulfate;decahydrate
Molecular Weight (g/mol) 322.195
Formula Weight 322.037
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.