missing translation for 'onlineSavingsMsg'
Learn More

Sodium sulphate, BAKER ANALYZED™, J.T. Baker™

Product Code. p-7220826 Shop All J.T.Baker Products
Click to view available options
Quantity:
500 g
1 kg
5 kg
Unit Size:
1kg
500g
5kg
This item is not returnable. View return policy

Product Code. 10456890

Brand: JT Baker 0313.0500

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Laboratory Reagent

Chemical Identifiers

CAS 7757-82-6
Molecular Formula Na2O4S
Molecular Weight (g/mol) 142.04
MDL Number MFCD00003504
InChI Key PMZURENOXWZQFD-UHFFFAOYSA-L
Synonym Sulphuric acid disodium salt , Bisodium sulphate , Disodium sulphate
IUPAC Name disodium sulfate
SMILES [Na+].[Na+].[O-]S([O-])(=O)=O

Specifications

Boiling Point 1429°C (1013 hPa)
Melting Point 888°C
pH 5.2 to 9.2 (5% Solution at 25°C)
Quantity 500 g
Assay Percent Range min. 99.0%
Loss on Ignition max. 0.5%
Packaging Glass bottle for solids
Density 2.7 g/cm³ (20°C)
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.