missing translation for 'onlineSavingsMsg'
Learn More

Sodium sulfite, anhydrous, 98%

Product Code. 11469056
Change view
Click to view available options
Quantity:
10,000 g
2500 g
500 g
Unit Size:
10000g
2500g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11469056 2500 g 2500g
11459056 500 g 500g
11479056 10,000 g 10000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11469056 Supplier Thermo Scientific Alfa Aesar Supplier No. A17933.0E
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 7757-83-7 | Na2O3S | 126.037 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
Sodium sulfite is used in the photography industry to protect developing solutions from oxidation, in the pulp and paper industry, in water treatment as an oxygen scavenger agent, as a desulfurizing and dechlorinating agent in the leather industry and as a bleaching agent in textile industry. It is used as a preservative to prevent dried fruit from discoloring. It finds application as a sulfonation and sulfomethylation agent in the chemical industry. It is involved in the production of sodium thiosulfate. Other applications include forth flotation ores, oil recovery and making dyes.

Solubility
Soluble in water and glycerol. Insoluble in alcohol, liquid chlorine and liquid ammonia.

Notes
Hygroscopic. Moisture and air sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with acids, strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7757-83-7
Molecular Formula Na2O3S
Molecular Weight (g/mol) 126.037
MDL Number MFCD00003503
InChI Key GEHJYWRUCIMESM-UHFFFAOYSA-L
Synonym sodium sulfite, disodium sulfite, sodium sulphite, sodium sulfite anhydrous, sulfurous acid, disodium salt, sulftech, natrium sulfurosum, sodiumsulfite, natriumsulfid, natriumsulfit
PubChem CID 24437
ChEBI CHEBI:86477
IUPAC Name disodium;sulfite
SMILES [O-]S(=O)[O-].[Na+].[Na+]

Specifications

Melting Point >500°C (decomposition)
Quantity 2500 g
Merck Index 14,8682
Solubility Information Soluble in water and glycerol. Insoluble in alcohol,liquid chlorine and liquid ammonia.
Formula Weight 126.04
Percent Purity 98%
Density 2.633
Chemical Name or Material Sodium sulfite, anhydrous

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.