missing translation for 'onlineSavingsMsg'
Learn More

Sodium Sulfide Nonahydrate, ≥98.0% Honeywell™ Fluka™

Product Code. 15628080 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
500 g
Packaging:
Poly bottle
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15628080 500 g Poly bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15628080 Supplier Honeywell-Fluka Supplier No. S2006500G

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 1313-84-4
Quantity 500 g
Molecular Formula H18Na2O9S
Linear Formula Na2S·9H2O
MDL Number MFCD00149184
UN Number UN1849
InChI Key ZGHLCBJZQLNUAZ-UHFFFAOYSA-N
SMILES O.O.O.O.O.O.O.O.O.[Na+].[Na+].[S--]
IUPAC Name disodium nonahydrate sulfanediide
Molecular Weight (g/mol) 240.17
Formula Weight 240.18g/mol
Percent Purity ≥98.0%
Packaging Poly bottle
Chemical Name or Material Sodium sulfide nonahydrate
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Sodium sulfide nonahydrate
Signal Word
  • Danger
Hazard Category
  • Substances and mixtures corrosive to metals Category 1
  • Acute toxicity Category 3
  • Skin corrosion/irritation Category 1 B
  • Serious eye damage/eye irritation Category 1
  • ACUTE AQUATIC Acute 1
Hazard Statement
  • H290-May be corrosive to metals.
  • H301+H311-Toxic if swallowed or in contact with skin
  • H314-Causes severe skin burns and eye damage.
  • H400-Very toxic to aquatic life.
  • EUH031-Contact with acids liberates toxic gas.
  • EUH071-Corrosive to the respiratory tract.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-[In case of inadequate ventilation] wear respiratory protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.