missing translation for 'onlineSavingsMsg'
Learn More

Sodium Sulfide Hydrate (Technical Grade), TRC

Product Code. 30138663
Change view
Click to view available options
Quantity:
10 g
50 g
100 g
Unit Size:
100g
10g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30138663 100 g 100g
30134094 50 g 50g
30011942 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30138663 Supplier Toronto Research Chemicals Supplier No. TRCS670985100G

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.
, 02 950 59 478

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 27610-45-3
Molecular Formula Na2S .xH2O
Molecular Weight (g/mol) 78.04
Synonym Disodium Sulfide Hydrate, Monosulfide Hydrate
IUPAC Name sodiosulfanylsodium
SMILES [Na]S[Na]

Specifications

Quantity 100 g
InChI Formula InChI=1S/2Na.S
Formula Weight 77.9516
Chemical Name or Material Sodium Sulfide Hydrate (Technical Grade)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.