missing translation for 'onlineSavingsMsg'
Learn More

Sodium sulfate purum p.a. anhydrous, Honeywell™

Product Code. 16605923
Change view
Click to view available options
50 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
16605923 50 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16605923 Supplier Fluka Supplier No. 71962S50KG
  • ONLINE EXCLUSIVE PRICE
    £1075.00
    £1042.00 / 50kg
    Save £33.00
    3% Off
 Restricted Item
Request Bulk or Custom Formats
Currently Unavailable: We have no information when this product will be available.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

Melting Point 884°C
CAS 7757-82-6
Color Colorless
CAS Max % 1
Quantity 50 kg
Molecular Formula Na2O4S
MDL Number MFCD00003504
InChI Key PMZURENOXWZQFD-UHFFFAOYSA-L
SMILES [Na+].[Na+].[O-]S([O-])(=O)=O
IUPAC Name disodium sulfate
Molecular Weight (g/mol) 142.04
Density 2.68 g/cm3 (20°C)
Chemical Name or Material Sodium sulfate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.