missing translation for 'onlineSavingsMsg'
Learn More

Sodium sulfate, Free-flowing, Reagent Grade, anhydrous, ≥99%, Solstice

Product Code. 15647110 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
500 g
1 kg
2.5 kg
5 kg
12 kg
Packaging:
Fiberboard Box
Poly Bottle
Unit Size:
12kg
1kg
2.5kg
500g
5kg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15647110 12 kg Fiberboard Box 12kg
15677110 500 g Poly Bottle 500g
15657110 1 kg Poly Bottle 1kg
15667110 2.5 kg Poly Bottle 2.5kg
15687110 5 kg Poly Bottle 5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 15647110 Supplier Honeywell-Fluka Supplier No. 79353112KG
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

Currently Unavailable: We have no information when this product will be available.
View alternative products
This item is not returnable. View return policy

Free-flowing, Reagent Grade, anhydrous,≥99%

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7757-82-6
Molecular Formula Na2O4S
Molecular Weight (g/mol) 142.04
MDL Number MFCD00003504
InChI Key PMZURENOXWZQFD-UHFFFAOYSA-L
Synonym sodium sulfate, disodium sulfate, salt cake, thenardite, sodium sulfate anhydrous, sodium sulphate, sodium sulfate, anhydrous, sulfuric acid disodium salt, disodium sulphate, sodium sulfate 2:1
PubChem CID 24436
ChEBI CHEBI:32149
IUPAC Name disodium;sulfate
SMILES [Na+].[Na+].[O-]S([O-])(=O)=O

Specifications

Melting Point 884°C (literature)
CAS Max % 100.0000%
Quantity 12 kg
Linear Formula Na2SO4
Formula Weight 142.04
Percent Purity >99%
Grade Reagent
Packaging Fiberboard Box
Density 2.68 g/mL at 25 °C(lit.)
Chemical Name or Material Sodium sulfate, Free-flowing, Reagent Grade, anhydrous, ≥99%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.